missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenol, ≥99.5%, Honeywell™ Fluka™

Product Code. p-7120096 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 15686940

Brand: Honeywell-Fluka 7356025G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 100-02-7
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00007331
InChI Key BTJIUGUIPKRLHP-UHFFFAOYSA-N
PubChem CID 980
ChEBI CHEBI:16836
IUPAC Name 4-nitrophenol
SMILES OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 116°C
Boiling Point 279°C
Linear Formula O2NC6H4OH
UN Number UN1663
Beilstein 1281877
Solubility Information 14.8g/L at 25°C
Formula Weight 139.11g/mol
Percent Purity ≥99.5%
Packaging Glass bottle
Vapor Pressure 0.6mmHg (120°C)
Density 1.479g/cm3
Color Yellow
pH 4.4
Quantity 25 g
Physical Form Crystalline
Chemical Name or Material 4-Nitrophenol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.