Learn More
4-Nitrobenzhydrazide, 98+%
Product Code. 11409156
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. 11409156
Brand: Thermo Scientific Alfa Aesar A18016.18
Request Bulk or Custom Format
CAS: 636-97-5 | C7H7N3O3 | 181.151 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 636-97-5
Molecular Formula C7H7N3O3
Molecular Weight (g/mol) 181.151
MDL Number MFCD00007604
InChI Key FKZXYJYTUSGIQE-UHFFFAOYSA-N
Synonym 4-nitrobenzhydrazide, p-nitrobenzhydrazide, 4-nitrobenzoylhydrazine, p-nitrobenzohydrazide, p-nitrobenzoylhydrazide, p-nitrobenzoic hydrazide, p-nitrobenzoic acid hydrazide, p-nitrobenzoyl hydrazine, 4-nitrobenzoic acid hydrazide, 4-nitrobenzoyl hydrazide
PubChem CID 3693744
ChEBI CHEBI:66928
IUPAC Name 4-nitrobenzohydrazide
SMILES C1=CC(=CC=C1C(=O)NN)[N+](=O)[O-]
Specifications
Melting Point ∼216°C (decomposition)
Quantity 50 g
Beilstein 519882
Formula Weight 181.15
Percent Purity ≥98%
Chemical Name or Material 4-Nitrobenzhydrazide