missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-3-(trifluoromethyl)benzenesulfonyl chloride, 97%

Product Code. 11365555
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11365555

Brand: Thermo Scientific Alfa Aesar L19721.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

CAS: 39234-83-8 | C7H3ClF3NO4S | 289.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39234-83-8
Molecular Formula C7H3ClF3NO4S
Molecular Weight (g/mol) 289.61
MDL Number MFCD03094387
InChI Key LJQUNSRHHHYXEW-UHFFFAOYSA-N
Synonym 4-nitro-3-trifluoromethyl benzenesulfonyl chloride, 4-nitro-3-trifluoromethyl benzenesulphonyl chloride, 4-nitro-3-trifluoromethyl benzenesulfonylchloride, 4-nitro-3-trifluoromethyl benzene-1-sulfonyl chloride, pubchem11722, acmc-1ail1, 5-chlorosulphonyl-2-nitrobenzotrifluoride, 4-nitro-3-trifluoromethyl phenylsulfonyl chloride, 4-chlorosulphonyl-2-trifluoromethyl nitrobenzene, 4-nitro-3-trifluoromethyl-benzenesulfonyl chloride
PubChem CID 2778595
IUPAC Name 4-nitro-3-(trifluoromethyl)benzenesulfonyl chloride
SMILES [O-][N+](=O)C1=C(C=C(C=C1)S(Cl)(=O)=O)C(F)(F)F

Specifications

Melting Point 63°C to 65°C
Quantity 1 g
UN Number UN3261
Beilstein 2880842
Sensitivity Moisture sensitive
Formula Weight 289.62
Percent Purity 97%
Chemical Name or Material 4-Nitro-3-(trifluoromethyl)benzenesulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.