Learn More
Alfa Aesar™ 4'-n-Propylacetophenone, 97%
Product Code. 11364474
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11364474 100 g 100g
11354474 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11364474 Supplier Alfa Aesar™ Supplier No. L01144.22
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 2932-65-2
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00041359
InChI Key ZNBVIYMIVFKTIW-UHFFFAOYSA-N
Synonym p-propylacetophenone, 1-4-propylphenyl ethan-1-one, 1-4-propylphenyl ethanone, 4'-propylacetophenone, ethanone, 1-4-propylphenyl, 4-n-propylacetophenone, 4'-n-propylacetophenone, 1-acetyl-4-propylbenzene, p-propylacetophenon, p-n-propylacetophenone
PubChem CID 76236
IUPAC Name 1-(4-propylphenyl)ethanone
SMILES CCCC1=CC=C(C=C1)C(C)=O
Specifications
Density 0.98
Boiling Point 251°C to 252°C
Refractive Index 1.5225
Quantity 100 g
Beilstein 1858604
Formula Weight 162.23
Percent Purity 97%
Chemical Name or Material 4'-n-Propylacetophenone