accessibility menu, dialog, popup

UserName

CH3CH3O

CAS RN 2932-65-2

IUPAC Name: 1-(4-propylphenyl)ethan-1-one
Synonyms: 1-(4-propylphenyl)ethanone
Molecular Weight (g/mol): 162.23
Molecular Formula: C11H14O
InChi Key: ZNBVIYMIVFKTIW-UHFFFAOYSA-N
SMILES: CCCC1=CC=C(C=C1)C(C)=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)