Learn More
4-n-Heptylaniline, 98%
Product Code. 11386889
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386889 5 g 5g
11376889 1 g 1g
11396889 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11386889 Supplier Thermo Scientific Alfa Aesar Supplier No. H26793.06
Request Bulk or Custom Format
CAS: 37529-27-4 | C13H21N | 191.32 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 37529-27-4
Molecular Formula C13H21N
Molecular Weight (g/mol) 191.32
MDL Number MFCD00007928
InChI Key BNEWZYZRLNNWNR-UHFFFAOYSA-N
Synonym p-heptylaniline, 4-n-heptylaniline, benzenamine, 4-heptyl, 4-heptylphenylamine, acmc-209ito, 4-heptylaniline
PubChem CID 142169
IUPAC Name 4-heptylaniline
SMILES CCCCCCCC1=CC=C(N)C=C1
Specifications
Density 0.91
Boiling Point 159°C (18 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.52
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 191.31
Percent Purity 98%
Chemical Name or Material 4-n-Heptylaniline