CAS RN 37529-27-4
CAS RN 37529-27-4
IUPAC Name:
4-heptylaniline
Synonyms:
4-heptylaniline
Molecular Weight (g/mol):
191.32
Molecular Formula:
C13H21N
InChi Key:
BNEWZYZRLNNWNR-UHFFFAOYSA-N
SMILES:
CCCCCCCC1=CC=C(N)C=C1
1
–
1
of
1
results
1
4-n-Heptylaniline, 98%
CAS: 37529-27-4 Molecular Formula: C13H21N Molecular Weight (g/mol): 191.32 MDL Number: MFCD00007928 InChI Key: BNEWZYZRLNNWNR-UHFFFAOYSA-N Synonym: p-heptylaniline,4-n-heptylaniline,benzenamine, 4-heptyl,4-heptylphenylamine,acmc-209ito,4-heptylaniline PubChem CID: 142169 IUPAC Name: 4-heptylaniline SMILES: CCCCCCCC1=CC=C(N)C=C1