missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Molecular weight: 288.34 g/mol
- Purity: >95%
- Application: Fluorescent substrate for esterases and lipases; also used for measuring cell proliferation and viability
- Composition: 100% 4-Methylumbelliferyl heptanoate
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 100% 4-Methylumbelliferyl heptanoate |
| Content And Storage | Store at -20°C. |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Substrate | 4-Methylumbelliferyl heptanoate |
| Target Molecule | Esterase, Lipase |
| Detection Method | Fluorescence |
| Formulation | 100% 4-Methylumbelliferyl heptanoate |
| Quantity | 100 mg |
| For Use With (Application) | Esterase activity measurement, Lipase activity measurement, Cell proliferation measurement, Cell viability measurement |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?