missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl Alpha-L-Iduronide (free acid), TRC

Product Code. 30136510
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136510 25 mg 25mg
30006020 5 mg 5mg
30008893 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136510 Supplier Toronto Research Chemicals Supplier No. TRCM33764325MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66966-09-4
Molecular Formula C16 H16 O9
Molecular Weight (g/mol) 352.29
Synonym α-L-Idopyranosiduronic acid, 4-methyl-2-oxo-2H-1-benzopyran-7-yl, 4-Methyl-2-oxo-2H-1-benzopyran-7-yl α-L-idopyranosiduronic acid, 4-Methylumbelliferyl α-L-idopyranosiduronic acid, 4-Methylumbelliferyl α-L-iduronide
IUPAC Name (2R,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxane-2-carboxylic acid
SMILES CC1=CC(=O)Oc2cc(O[C@@H]3O[C@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O)ccc12

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H16O9/c1-6-4-10(17)24-9-5-7(2-3-8(6)9)23-16-13(20)11(18)12(19)14(25-16)15(21)22/h2-5,11-14,16,18-20H,1H3,(H,21,22)/t11-,12-,13+,14+,16+/m0/s1
Formula Weight 352.0794
Percent Purity >95%
Chemical Name or Material 4-Methylumbelliferyl alpha-L-Iduronide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.