CAS RN 66966-09-4
CAS RN 66966-09-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Methylumbelliferyl Alpha-L-Iduronide (free acid), TRC
CAS: 66966-09-4 Molecular Formula: C16 H16 O9 Molecular Weight (g/mol): 352.29 Synonym: α-L-Idopyranosiduronic acid, 4-methyl-2-oxo-2H-1-benzopyran-7-yl,4-Methyl-2-oxo-2H-1-benzopyran-7-yl α-L-idopyranosiduronic acid,4-Methylumbelliferyl α-L-idopyranosiduronic acid,4-Methylumbelliferyl α-L-iduronide IUPAC Name: (2R,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxane-2-carboxylic acid SMILES: CC1=CC(=O)Oc2cc(O[C@@H]3O[C@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O)ccc12