Learn More
4-Methyl-3-[[(2RS)-2-(propylamino)propanoyl]amino]thiophene-2-carboxylic Acid Hydrochloride (Articaine Acid Hydrochloride), Mikromol™
Description
4-Methyl-3-[[(2RS)-2-(propylamino)propanoyl]amino]thiophene-2-carboxylic Acid Hydrochloride (Articaine Acid Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | Articaine Acid Hydrochloride |
| CAS | 2984195-05-1 |
| Analyte or Component Names | Articaine Acid Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H18 N2 O3 S . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | 4-Methyl-3-[[(2RS)-2-(propylamino)propanoyl]amino]thiophene-2-carboxylic Acid, Articaine Hydrochloride Imp. B (EP), Articaine Acid, Articainic acid, 4-Methyl-3-[[1-oxo-2-(propylamino)propyl]amino]-2-thiophenecarboxylic acid, Articaine USP Related Compound B, Articaine USP RC B, 4-Methyl-3-[[(2RS)-2-(propylamino)propanoyl]amino]thiophene-2-carboxylic Acid Hydrochloride, Articaine Hydrochloride Imp. B (EP) as Hydrochloride, Articaine Acid Hydrochloride, Articaine Related Compound B as Hydrochloride, Articaine Hydrochloride Impurity B as Hydrochloride, Articaine Impurity B as Hydrochloride |
| InChI Formula | InChI=1S/C12H18N2O3S.ClH/c1-4-5-13-8(3)11(15)14-9-7(2)6-18-10(9)12(16)17;/h6,8,13H,4-5H2,1-3H3,(H,14,15)(H,16,17);1H |
| SMILES | Cl.CCCNC(C)C(=O)Nc1c(C)csc1C(=O)O |
| IUPAC Name | 4-methyl-3-[2-(propylamino)propanoylamino]thiophene-2-carboxylic acid;hydrochloride |
| Show More |