missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-pentyn-3-ol, 97%

Product Code. 11347395
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347395 10 g 10g
11357395 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347395 Supplier Thermo Scientific Alfa Aesar Supplier No. L05318.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 565-68-4 | C6H10O | 98.145 g/mol

4-Methyl-1-pentyn-3-ol, is used as standard Pharmaceutical intermediate and also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methyl-1-pentyn-3-ol, is used as standard Pharmaceutical intermediate and also used in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565-68-4
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00039847
InChI Key UTIFIONYBLSHIL-UHFFFAOYSA-N
Synonym 4-methyl-1-pentyn-3-ol, 4-methylpentyn-3-ol, 1-pentyn-3-ol, 4-methyl, 1-isopropylpropargyl alcohol, 4-methyl-1-pentyne-3-ol, 2-methyl-4-pentyn-3-ol, 4-methyl-pent-1-yn-3-ol, acmc-1as78, 4-methyl-1-pentyn-3-ol 10g
PubChem CID 98279
IUPAC Name 4-methylpent-1-yn-3-ol
SMILES CC(C)C(C#C)O

Specifications

Density 0.929
Boiling Point 133°C to 134°C
Flash Point 41°C (105°F)
Odor Recognizable
Refractive Index 1.436
Quantity 10 g
UN Number UN1987
Beilstein 1698762
Solubility Information Slightly soluble in water.
Formula Weight 98.15
Percent Purity 97%
Chemical Name or Material 4-Methyl-1-pentyn-3-ol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.