accessibility menu, dialog, popup

UserName

CH3H3COHHC

CAS RN 565-68-4

IUPAC Name: 4-methylpent-1-yn-3-ol
Synonyms: 4-methylpent-1-yn-3-ol
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: UTIFIONYBLSHIL-UHFFFAOYNA-N
SMILES: CC(C)C(O)C#C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)