Learn More
4-Methoxy-3-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™
Product Code. 11392459
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11392459 5 g 5g
11382459 1 g 1g
2 options
Product Code. 11392459
Brand: Thermo Scientific Alfa Aesar H26606.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
As organic intermediate, pharmaceutical intermediate. In chemical synthesis.
Solubility
Sparingly Soluble (0.16 g/L) (25°C).
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 261951-89-7
Molecular Formula C9H8BrF3O
Molecular Weight (g/mol) 269.061
MDL Number MFCD01631578
InChI Key FZICFAROPYFJMF-UHFFFAOYSA-N
Synonym 4-methoxy-3-trifluoromethyl benzyl bromide, 4-bromomethyl-1-methoxy-2-trifluoromethyl benzene, benzene, 4-bromomethyl-1-methoxy-2-trifluoromethyl, 4-bromomethyl-1-methoxy-2-trifluoromethyl-benzene, 4-bromomethyl-2-trifluoromethyl-1-methoxybenzene, acmc-20amfh, pubchem4655, 4-methoxy-3-trifluoromethyl benzylbromide
PubChem CID 2775302
IUPAC Name 4-(bromomethyl)-1-methoxy-2-(trifluoromethyl)benzene
SMILES COC1=C(C=C(C=C1)CBr)C(F)(F)F
Specifications
Melting Point 75°C to 79°C
Quantity 5 g
UN Number UN3261
Solubility Information Sparingly Soluble (0.16g/L) (25°C).
Formula Weight 269.06
Percent Purity 97%
Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)benzyl bromide