missing translation for 'onlineSavingsMsg'
Learn More

(4-Isopropoxyphenyl)hydrazine Hydrochloride, TRC

Product Code. 30391821
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391821 250 mg 250mg
30394306 1 g 1g
30402092 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391821 Supplier Toronto Research Chemicals Supplier No. TRCI918323250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 104033-62-7
Molecular Formula C9H14N2O . HCl
Molecular Weight (g/mol) 202.68
Synonym N-(2,5-Diethoxyphenyl)benzamide, 2',5'-Diethoxybenzanilide, NSC 163154
IUPAC Name (4-isopropoxyphenyl)hydrazine hydrochloride
SMILES CC(C)OC1=CC=C(C=C1)NN.Cl

Specifica

Quantity 250 mg
InChI Formula InChI=1S/C9H14N2O.ClH/c1-7(2)12-9-5-3-8(11-10)4-6-9;/h3-7,11H,10H2,1-2H3;1H
Formula Weight 202.09
Chemical Name or Material (4-Isopropoxyphenyl)hydrazine Hydrochloride
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.