missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-(trifluoromethoxy)benzaldehyde, 97%, Thermo Scientific™

Product Code. 11326729
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11326729 5 g 5g
11316729 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11326729 Supplier Thermo Scientific Alfa Aesar Supplier No. H26256.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86256-48-6
Molecular Formula C8H4F4O2
Molecular Weight (g/mol) 208.112
MDL Number MFCD06660159
InChI Key CGOFJJTXLIWONB-UHFFFAOYSA-N
Synonym 4-fluoro-3-trifluoromethoxy benzaldehyde, 5-formyl-alpha,alpha,alpha,2-tetrafluoroanisole, pubchem10075, 4-fluoro-3-trifluoromethoxy-benzaldehyde, 4-fluoranyl-3-trifluoromethyloxy benzaldehyde, benzaldehyde, 4-fluoro-3-trifluoromethoxy
PubChem CID 17750776
IUPAC Name 4-fluoro-3-(trifluoromethoxy)benzaldehyde
SMILES C1=CC(=C(C=C1C=O)OC(F)(F)F)F

Specifications

Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 208.11
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-(trifluoromethoxy)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.