accessibility menu, dialog, popup

UserName

FOOFFF

CAS RN 86256-48-6

IUPAC Name: 4-fluoro-3-(trifluoromethoxy)benzaldehyde
Synonyms: 4-fluoro-3-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol): 208.11
Molecular Formula: C8H4F4O2
InChi Key: CGOFJJTXLIWONB-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=O)C=C1OC(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results