Learn More
4-Ethoxy-2-fluorobenzoic acid, 97%, Thermo Scientific™
Product Code. 15467357
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15467357 5 g 5g
15457357 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15467357 Supplier Thermo Scientific Alfa Aesar Supplier No. H34414.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1206593-29-4
Molecular Formula C9H9FO3
Molecular Weight (g/mol) 184.166
MDL Number MFCD13196593
InChI Key WVXNAAKJFTYOCN-UHFFFAOYSA-N
PubChem CID 19978989
IUPAC Name 4-ethoxy-2-fluorobenzoic acid
SMILES CCOC1=CC(=C(C=C1)C(=O)O)F
Specifications
Quantity 5 g
Formula Weight 184.17
Percent Purity 97%
Chemical Name or Material 4-Ethoxy-2-fluorobenzoic acid