accessibility menu, dialog, popup

UserName

CH3OOHOF

CAS RN 1206593-29-4

IUPAC Name: 4-ethoxy-2-fluorobenzoic acid
Synonyms: 4-ethoxy-2-fluorobenzoic acid
Molecular Weight (g/mol): 184.17
Molecular Formula: C9H9FO3
InChi Key: WVXNAAKJFTYOCN-UHFFFAOYSA-N
SMILES: CCOC1=CC(F)=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results