Learn More
4-Dimethylaminobutyric acid hydrochloride, 98%
Product Code. 11437957
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11437957 25 g 25g
11427957 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11437957 Supplier Thermo Scientific Alfa Aesar Supplier No. A16610.14
 Restricted Item
In Stock
Add to basket
CAS: 69954-66-1 | C6H14ClNO2 | 167.633 g/mol
4-Dimethylaminobutyric acid hydrochloride is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Dimethylaminobutyric acid hydrochloride is used as pharmaceutical intermediates.
Notes
Hygroscopic. Protect from humidity and water. Store under dry inert gas. Incompatible with water and oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 69954-66-1
Molecular Formula C6H14ClNO2
Molecular Weight (g/mol) 167.633
MDL Number MFCD00012613
InChI Key RDTALXUBMCLWBB-UHFFFAOYSA-N
Synonym 4-dimethylamino butanoic acid hydrochloride, 4-dimethylaminobutyric acid hydrochloride, 4-dimethylamino butyric acid hydrochloride, butanoic acid, 4-dimethylamino-, hydrochloride, 4-dimethylamino butanoic acid hcl, 4-dimethylamino butyric acid hcl salt, 4-dimethylamino-butyric acid hydrochloride, 4-dimethylamino-butyric acid hydrochloride salt
PubChem CID 12691640
IUPAC Name 4-(dimethylamino)butanoic acid;hydrochloride
SMILES CN(C)CCCC(=O)O.Cl
Specifications
Melting Point 153°C to 155°C
Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 167.64
Percent Purity 98%
Chemical Name or Material 4-Dimethylaminobutyric acid hydrochloride
compliance-icons
Product Identifier
  • 4-Dimethylaminobutyric acid, hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Butanoic acid, 4-(dimethylamino)-, hydrochloride