CAS RN 69954-66-1
IUPAC Name:
hydrogen 4-(dimethylamino)butanoic acid chloride
Synonyms:
hydrogen 4-(dimethylamino)butanoic acid chloride
Molecular Weight (g/mol):
167.63
Molecular Formula:
C6H14ClNO2
InChi Key:
RDTALXUBMCLWBB-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CN(C)CCCC(O)=O
1
–
1
of
1
results
1
4-Dimethylaminobutyric acid hydrochloride, 98%
CAS: 69954-66-1 Molecular Formula: C6H14ClNO2 Molecular Weight (g/mol): 167.633 MDL Number: MFCD00012613 InChI Key: RDTALXUBMCLWBB-UHFFFAOYSA-N Synonym: 4-dimethylamino butanoic acid hydrochloride,4-dimethylaminobutyric acid hydrochloride,4-dimethylamino butyric acid hydrochloride,butanoic acid, 4-dimethylamino-, hydrochloride,4-dimethylamino butanoic acid hcl,4-dimethylamino butyric acid hcl salt,4-dimethylamino-butyric acid hydrochloride,4-dimethylamino-butyric acid hydrochloride salt PubChem CID: 12691640 IUPAC Name: 4-(dimethylamino)butanoic acid;hydrochloride SMILES: CN(C)CCCC(=O)O.Cl