Learn More
4'-Demethylepipodophyllotoxin 9-(beta-D-Glucopyranoside) (Lignan P), Mikromol™
Description
4'-Demethylepipodophyllotoxin 9-(beta-D-Glucopyranoside) (Lignan P)
Specifications
Specifications
| Chemical Name or Material | Lignan P |
| CAS | 23363-35-1 |
| Analyte or Component Names | Lignan P |
| Shipping Condition | Room Temperature |
| Molecular Formula | C27 H30 O13 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one, 9-(β-D-glucopyranosyloxy)-5,8,8a,9-tetrahydro-5-(4-hydroxy-3,5-dimethoxyphenyl)-, (5R,5aR,8aR,9S)-, Epipodophyllotoxin, 4'-demethyl-, 9-β-D-glucopyranoside (8CI), Furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one, 9-(β-D-glucopyranosyloxy)-5,8,8a,9-tetrahydro-5-(4-hydroxy-3,5-dimethoxyphenyl)-, [5R-(5α,5aβ,8aα,9β)]-, (5R,5aR,8aR,9S)-9-(β-D-Glucopyranosyloxy)-5,8,8a,9-tetrahydro-5-(4-hydroxy-3,5-dimethoxyphenyl)furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one, Glucopyranoside, 4'-demethylepipodophyllotoxin, β-D- (8CI), 4-Demethylepipodophyllotoxin 7'-O-β-D-glucopyranoside, 4'-Demethylepipodophyllotoxin glucoside, 4'-Demethylepipodophyllotoxin β-D-glucopyranoside, Lignan P, Pptoxin II, 4'-Demethylepipodophyllotoxin 9-(β-D-glucopyranoside), (5R,5aR,8aR,9S)-9-(β-D-Glucopyranosyloxy)-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,8,8a,9-tetrahydro[2]benzofuro-[5,6-f][1,3]benzodioxol-6(5aH)-one, Etoposide Impurity D |
| InChI Formula | InChI=1S/C27H30O13/c1-34-16-3-10(4-17(35-2)21(16)29)19-11-5-14-15(38-9-37-14)6-12(11)25(13-8-36-26(33)20(13)19)40-27-24(32)23(31)22(30)18(7-28)39-27/h3-6,13,18-20,22-25,27-32H,7-9H2,1-2H3/t13-,18+,19+,20-,22+,23-,24+,25+,27-/m0/s1 |
| SMILES | COc1cc(cc(OC)c1O)[C@H]2[C@@H]3[C@H](COC3=O)[C@H](O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)c5cc6OCOc6cc25 |
| IUPAC Name | (5S,5aR,8aR,9R)-9-(4-hydroxy-3,5-dimethoxyphenyl)-5-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-5a,6,8a,9-tetrahydro-5H-[2]benzofuro[5,6-f][1,3]benzodioxol-8-one |
| Show More |