missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)-1,2-diphenylethane, 98%

Product Code. 11490728
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11490728

Brand: Thermo Scientific Alfa Aesar B25222.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 80676-35-3 | C15H15Cl | 230.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80676-35-3
Molecular Formula C15H15Cl
Molecular Weight (g/mol) 230.74
MDL Number MFCD02093918
InChI Key KPTSLALCCGBCOB-UHFFFAOYSA-N
Synonym 4-chloromethyl dibenzyl, 1-chloromethyl-4-phenethylbenzene, 4-chloromethyl-1,2-diphenylethane, 1-chloromethyl-4-2-phenylethyl benzene, 4-chloromethyl bibenzyl, 4-phenethylbenzyl chloride
PubChem CID 2733990
IUPAC Name 1-(chloromethyl)-4-(2-phenylethyl)benzene
SMILES ClCC1=CC=C(CCC2=CC=CC=C2)C=C1

Specifications

Melting Point 38°C to 42°C
Quantity 25 g
UN Number UN1759
Formula Weight 230.73
Percent Purity 98%
Chemical Name or Material 4-(Chloromethyl)-1,2-diphenylethane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.