accessibility menu, dialog, popup

UserName

CAS RN 80676-35-3

Cl

CAS RN 80676-35-3

IUPAC Name: 1-(chloromethyl)-4-(2-phenylethyl)benzene
Synonyms: 1-(chloromethyl)-4-(2-phenylethyl)benzene
Molecular Weight (g/mol): 230.74
Molecular Formula: C15H15Cl
InChi Key: KPTSLALCCGBCOB-UHFFFAOYSA-N
SMILES: ClCC1=CC=C(CCC2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results