Learn More
4-(Chloromethyl)-1,2-diphenylethane, 98%
Product Code. 11470728
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11470728
Brand: Thermo Scientific Alfa Aesar B25222.03
Request Bulk or Custom Format
CAS: 80676-35-3 | C15H15Cl | 230.74 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 80676-35-3
Molecular Formula C15H15Cl
Molecular Weight (g/mol) 230.74
MDL Number MFCD02093918
InChI Key KPTSLALCCGBCOB-UHFFFAOYSA-N
Synonym 4-chloromethyl dibenzyl, 1-chloromethyl-4-phenethylbenzene, 4-chloromethyl-1,2-diphenylethane, 1-chloromethyl-4-2-phenylethyl benzene, 4-chloromethyl bibenzyl, 4-phenethylbenzyl chloride
PubChem CID 2733990
IUPAC Name 1-(chloromethyl)-4-(2-phenylethyl)benzene
SMILES ClCC1=CC=C(CCC2=CC=CC=C2)C=C1
Specifications
Melting Point 38°C to 42°C
Quantity 1 g
UN Number UN1759
Formula Weight 230.73
Percent Purity 98%
Chemical Name or Material 4-(Chloromethyl)-1,2-diphenylethane