Learn More
4-Chloro-7-methoxy-6-quinolinecarboxamide, TRC
Product Code. 30367655
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367655 250 mg 250mg
30361916 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30367655 Supplier Toronto Research Chemicals Supplier No. TRCC369515250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 417721-36-9
Molecular Formula C11H9ClN2O2
Molecular Weight (g/mol) 236.65
Synonym 4-Chloro-7-methoxyquinoline-6-carboxamide
IUPAC Name 4-chloro-7-methoxyquinoline-6-carboxamide
SMILES COc1cc2nccc(Cl)c2cc1C(=N)O
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C11H9ClN2O2/c1-16-10-5-9-6(4-7(10)11(13)15)8(12)2-3-14-9/h2-5H,1H3,(H2,13,15)
Formula Weight 236.0353
Chemical Name or Material 4-Chloro-7-methoxy-6-quinolinecarboxamide