CAS RN 417721-36-9
CAS RN 417721-36-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Chloro-7-methoxy-6-quinolinecarboxamide, TRC
CAS: 417721-36-9 Molecular Formula: C11H9ClN2O2 Molecular Weight (g/mol): 236.65 Synonym: 4-Chloro-7-methoxyquinoline-6-carboxamide IUPAC Name: 4-chloro-7-methoxyquinoline-6-carboxamide SMILES: COc1cc2nccc(Cl)c2cc1C(=N)O