missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluorobenzoic acid, 98%

Product Code. 11460848
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460848 25 g 25g
11440848 1 g 1g
11450848 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460848 Supplier Thermo Scientific Alfa Aesar Supplier No. B25277.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

CAS: 446-30-0 | C7H4ClFO2 | 174.555 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-30-0
Molecular Formula C7H4ClFO2
Molecular Weight (g/mol) 174.555
MDL Number MFCD00042468
InChI Key ZLPXBWMVZANJJQ-UHFFFAOYSA-N
Synonym 2-fluoro-4-chlorobenzoic acid, benzoic acid, 4-chloro-2-fluoro, 4-chloro-2-fluorobenzoicacid, 4-chloro-2-fluoro-benzoic acid, pubchem1374, acmc-209jz4, ksc237m4j, rarechem al bo 0755, 4-chloro-2-fluoro benzoic acid, timtec-bb sbb003797
PubChem CID 99152
IUPAC Name 4-chloro-2-fluorobenzoic acid
SMILES C1=CC(=C(C=C1Cl)F)C(=O)O

Specifications

Melting Point 203°C to 206°C
Quantity 25 g
Formula Weight 174.56
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-fluorobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.