accessibility menu, dialog, popup

UserName

CAS RN 446-30-0

HOOClF

CAS RN 446-30-0

IUPAC Name: 4-chloro-2-fluorobenzoic acid
Synonyms: 4-chloro-2-fluorobenzoic acid
Molecular Weight (g/mol): 174.56
Molecular Formula: C7H4ClFO2
InChi Key: ZLPXBWMVZANJJQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(Cl)C=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results