missing translation for 'onlineSavingsMsg'
Learn More

4-(Benzyl(methyl)amino)-N-methoxy-N-methylbenzamide, TRC

Product Code. 30335689
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 30335689

Brand: Toronto Research Chemicals TRCB28785510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Benzyl(methyl)amino)-N-methoxy-N-methylbenzamide
Analyte or Component Names 4-(Benzyl(methyl)amino)-N-methoxy-N-methylbenzamide
Shipping Condition Room Temperature
Molecular Formula C17 H20 N2 O2
InChI Formula InChI=1S/C17H20N2O2/c1-18(13-14-7-5-4-6-8-14)16-11-9-15(10-12-16)17(20)19(2)21-3/h4-12H,13H2,1-3H3
Solution Type Neat
SMILES CON(C)C(=O)c1ccc(cc1)N(C)Cc2ccccc2
IUPAC Name 4-[benzyl(methyl)amino]-N-methoxy-N-methylbenzamide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 284.353
Quantity 10 mg
Formula Weight 284.152
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.