missing translation for 'onlineSavingsMsg'
Learn More

4-Azidobenzaldehyde, TRC

Product Code. 30134702
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134702 2.5 g 2.5g
30005946 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134702 Supplier Toronto Research Chemicals Supplier No. TRCA8220402.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24173-36-2
Molecular Formula C7 H5 N3 O
Molecular Weight (g/mol) 147.13
Synonym p-Azidobenzaldehyde, p-Azido-benzaldehyde
IUPAC Name 4-azidobenzaldehyde
SMILES [N-]=[N+]=Nc1ccc(C=O)cc1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C7H5N3O/c8-10-9-7-3-1-6(5-11)2-4-7/h1-5H
Formula Weight 147.0433
Percent Purity >95%
Chemical Name or Material 4-Azidobenzaldehyde
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.