CAS RN 24173-36-2
CAS RN 24173-36-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Azidobenzaldehyde, TRC
CAS: 24173-36-2 Molecular Formula: C7 H5 N3 O Molecular Weight (g/mol): 147.13 Synonym: p-Azidobenzaldehyde,p-Azido-benzaldehyde IUPAC Name: 4-azidobenzaldehyde SMILES: [N-]=[N+]=Nc1ccc(C=O)cc1