Learn More
4-Androsten-3-one-17beta-carboxylic Acid, Mikromol™
Description
4-Androsten-3-one-17beta-carboxylic Acid, Mikromol
Specifications
Specifications
| Chemical Name or Material | 4-Androsten-3-one-17beta-carboxylic Acid |
| CAS | 302-97-6 |
| Analyte or Component Names | 4-Androsten-3-one-17beta-carboxylic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H28 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | (17β)-3-Oxo-androst-4-ene-17-carboxylic Acid, Androst-4-ene-17β-carboxylic acid, 3-oxo- (6CI,7CI,8CI), 17β-Carboxyandrost-4-en-3-one, 3-Carbonyl-4-androstene-17β-carboxylic acid, 3-Oxoandrost-4-ene-17β-carboxylic acid, AD-Acid, AL 3793, CC 14837, Etienic acid, L 552.803, NSC 226121, SKF 106224, Testosterone-17β-carboxylic acid, 4-Androsten-3-one-17β-carboxylic Acid, 3-Oxo-4-androstene-17β-carboxylic Acid, SKF-106224, Dutasteride related |
| InChI Formula | InChI=1S/C20H28O3/c1-19-9-7-13(21)11-12(19)3-4-14-15-5-6-17(18(22)23)20(15,2)10-8-16(14)19/h11,14-17H,3-10H2,1-2H3,(H,22,23)/t14-,15-,16-,17+,19-,20-/m0/s1 |
| Solution Type | Neat |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2C(=O)O |
| Show More |