missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Molecular weight: 211.09 Da
- Purity: >99%
- Physical state: Solid
- Application: Substrate for electrochemical alkaline phosphatase measurement
- Storage buffer: 100% sodium (4-aminophenyl) hydrogen phosphate
- Storage temperature: -20°C
- Shipping temperature: Ambient (can ship with ice)
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 100% sodium (4-aminophenyl) hydrogen phosphate |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Reactivity | Alkaline Phosphatase |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Substrate | 4-Aminophenyl phosphate |
| Target Molecule | Alkaline Phosphatase |
| Formulation | 100% sodium (4-aminophenyl) hydrogen phosphate |
| Quantity | 10 mg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?