missing translation for 'onlineSavingsMsg'
Learn More

4-Aminomethyl-1-(n-butyl)piperidine, 97%, Thermo Scientific™

Product Code. 11459690
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11459690

Brand: Thermo Scientific Alfa Aesar H50947.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65017-57-4
Molecular Formula C10H22N2
Molecular Weight (g/mol) 170.30
MDL Number MFCD02684339,MFCD08060490
InChI Key SXSSVNBOMMHRRB-UHFFFAOYSA-N
Synonym 4-aminomethyl-1-n-butylpiperidine, 4-aminomethyl-1-n-butyl piperidine, 1-butylpiperidin-4-yl methanamine, 1-n-butyl-4-piperidinemethylamine, 1-butyl-4-piperidyl methanamine, 4ambppr, acmc-1bc4b, 4-piperidinemethanamine,1-butyl, 1-butyl-4-piperidyl methylamine
PubChem CID 10442133
IUPAC Name (1-butylpiperidin-4-yl)methanamine
SMILES CCCCN1CCC(CN)CC1

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 170.3
Percent Purity 97%
Chemical Name or Material 4-Aminomethyl-1-(n-butyl)piperidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.