accessibility menu, dialog, popup

UserName

CAS RN 65017-57-4

H3CNNH2

CAS RN 65017-57-4

IUPAC Name: 1-(1-butylpiperidin-4-yl)methanamine
Synonyms: 1-(1-butylpiperidin-4-yl)methanamine
Molecular Weight (g/mol): 170.3
Molecular Formula: C10H22N2
InChi Key: SXSSVNBOMMHRRB-UHFFFAOYSA-N
SMILES: CCCCN1CCC(CN)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results