Learn More
4-(Acetylamino)-2-hydroxybenzoic Acid, Mikromol™
Description
4-(Acetylamino)-2-hydroxybenzoic Acid
Specifications
Specifications
| Chemical Name or Material | 4-(Acetylamino)-2-hydroxybenzoic Acid |
| CAS | 50-86-2 |
| Analyte or Component Names | 4-(Acetylamino)-2-hydroxybenzoic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H9 N O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 4-(Acetylamino)-2-hydroxybenzoic Acid, Metoclopramide Imp. H (EP), Metoclopramide Hydrochloride Monohydrate Impurity H, Metoclopramide Impurity H, Metoclopramide Impurity H |
| InChI Formula | InChI=1S/C9H9NO4/c1-5(11)10-6-2-3-7(9(13)14)8(12)4-6/h2-4,12H,1H3,(H,10,11)(H,13,14) |
| SMILES | CC(=O)Nc1ccc(C(=O)O)c(O)c1 |
| IUPAC Name | 4-acetamido-2-hydroxybenzoic acid |
| Show More |