Learn More
4-(5-Methyl-3-phenylisoxazol-4-yl)benzenesulfonic Acid Dihydrate (Valdecoxib Sulfonic Acid Dihydrate), Mikromol™
Description
4-(5-Methyl-3-phenylisoxazol-4-yl)benzenesulfonic Acid Dihydrate (Valdecoxib Sulfonic Acid Dihydrate)
Specifications
Specifications
| Chemical Name or Material | 4-(5-Methyl-3-phenylisoxazol-4-yl)benzenesulfonic Acid Dihydrate |
| CAS | 2987295-53-2 |
| Analyte or Component Names | 4-(5-Methyl-3-phenylisoxazol-4-yl)benzenesulfonic Acid Dihydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H13 N O4 S . 2 H2 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4-(5-Methyl-3-phenylisoxazol-4-yl)benzenesulfonic Acid Dihydrate |
| InChI Formula | InChI=1S/C16H13NO4S.2H2O/c1-11-15(12-7-9-14(10-8-12)22(18,19)20)16(17-21-11)13-5-3-2-4-6-13;;/h2-10H,1H3,(H,18,19,20);2*1H2 |
| SMILES | O.O.Cc1onc(c2ccccc2)c1c3ccc(cc3)S(=O)(=O)O |
| IUPAC Name | 4-(5-methyl-3-phenyl-1,2-oxazol-4-yl)benzenesulfonic acid;dihydrate |
| Show More |