Learn More
4,5-Dibromoveratrole, 98+%
Product Code. 11301477
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11301477 10 g 10g
11311477 50 g 50g
11321477 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11301477 Supplier Thermo Scientific Alfa Aesar Supplier No. L01999.09
Request Bulk or Custom Format
CAS: 37895-73-1 | C8H8Br2O2 | 295.958 g/mol
4,5-Dibromoveratrole is used as chemical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,5-Dibromoveratrole is used as chemical intermediates.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with Strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 37895-73-1
Molecular Formula C8H8Br2O2
Molecular Weight (g/mol) 295.958
MDL Number MFCD00014894
InChI Key ZYCLQXMMFJREPJ-UHFFFAOYSA-N
Synonym 4,5-dibromoveratrole, 4,5-dibromoveratrol, benzene, 1,2-dibromo-4,5-dimethoxy, acmc-1aeu5, amth147, 1.2-dibromo-4.5-dimethoxy-benzene, 1,2-bis bromanyl-4,5-dimethoxy-benzene
PubChem CID 2758033
IUPAC Name 1,2-dibromo-4,5-dimethoxybenzene
SMILES COC1=CC(=C(C=C1OC)Br)Br
Specifications
Melting Point 90°C to 92°C
Quantity 10 g
Beilstein 1953148
Solubility Information Insoluble in water.
Formula Weight 295.96
Percent Purity ≥98%
Chemical Name or Material 4,5-Dibromoveratrole