CAS RN 37895-73-1
CAS RN 37895-73-1
IUPAC Name:
1,2-dibromo-4,5-dimethoxybenzene
Synonyms:
1,2-dibromo-4,5-dimethoxybenzene
Molecular Weight (g/mol):
295.96
Molecular Formula:
C8H8Br2O2
InChi Key:
ZYCLQXMMFJREPJ-UHFFFAOYSA-N
SMILES:
COC1=CC(Br)=C(Br)C=C1OC
1
–
1
of
1
results
1
4,5-Dibromoveratrole, 98+%
CAS: 37895-73-1 Molecular Formula: C8H8Br2O2 Molecular Weight (g/mol): 295.958 MDL Number: MFCD00014894 InChI Key: ZYCLQXMMFJREPJ-UHFFFAOYSA-N Synonym: 4,5-dibromoveratrole,4,5-dibromoveratrol,benzene, 1,2-dibromo-4,5-dimethoxy,acmc-1aeu5,amth147,1.2-dibromo-4.5-dimethoxy-benzene,1,2-bis bromanyl-4,5-dimethoxy-benzene PubChem CID: 2758033 IUPAC Name: 1,2-dibromo-4,5-dimethoxybenzene SMILES: COC1=CC(=C(C=C1OC)Br)Br