missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methoxyphenyl)-2-butanone, 98%, Thermo Scientific™

Product Code. 10237660 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10237660 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10237660 Supplier Thermo Scientific Acros Supplier No. 313980500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-20-1
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00008791
InChI Key PCBSXBYCASFXTM-UHFFFAOYSA-N
Synonym 4-4-methoxyphenyl-2-butanone, anisylacetone, 4-4-methoxyphenyl butan-2-one, 4-methoxybenzylacetone, raspberry ketone methyl ether, 2-butanone, 4-4-methoxyphenyl, anisylacetone, p, 4-p-methoxyphenyl-2-butanone, methyl oxanone, 1-4-methoxyphenyl-3-butanone
PubChem CID 61007
IUPAC Name 4-(4-methoxyphenyl)butan-2-one
SMILES COC1=CC=C(CCC(C)=O)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.04g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3OC6H4CHCHCOCH3
Refractive Index 1.5190 to 1.5210 (20°C, 589nm)
Quantity 50 g
Formula Weight 178.23
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 4-(4-Methoxyphenyl)-2-butanone, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.