Learn More
4,4'-Diaminobenzanilide, 98%
Product Code. 11440547
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. 11440547
Brand: Thermo Scientific Alfa Aesar B22419.09
Request Bulk or Custom Format
CAS: 785-30-8 | C13H13N3O | 227.27 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 785-30-8
Molecular Formula C13H13N3O
Molecular Weight (g/mol) 227.27
MDL Number MFCD00025361
InChI Key XPAQFJJCWGSXGJ-UHFFFAOYSA-N
Synonym 4,4'-diaminobenzanilide, 4-amino-n-4-aminophenyl benzamide, benzamide, 4-amino-n-4-aminophenyl, 4-aminobenzoyl-4'-aminoanilide, p,p'-diaminobenzanilide, 4,4'-diamino benzanilide, unii-00x4jm89uh, benzanilide, 4,4'-diamino, 4-aminophenyl-n-4-aminophenyl carboxamide, ccris 8916
PubChem CID 69917
IUPAC Name 4-amino-N-(4-aminophenyl)benzamide
SMILES NC1=CC=C(NC(=O)C2=CC=C(N)C=C2)C=C1
Specifications
Melting Point 200°C to 202°C
Quantity 10 g
Beilstein 2215469
Formula Weight 227.27
Percent Purity 98%
Chemical Name or Material 4,4'-Diaminobenzanilide