Learn More
[4-(4-Aminophenoxy)(2-pyridyl)]-N-methylcarboxamide, TRC
Product Code. 30137677
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137677 5 g 5g
17941405 1 g 1g
17941185 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137677 Supplier Toronto Research Chemicals Supplier No. TRCA6189405G
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 284462-37-9
Molecular Formula C13 H13 N3 O2
Molecular Weight (g/mol) 243.26
Synonym 2-Pyridinecarboxamide, 4-(4-aminophenoxy)-N-methyl-, 4-(4-Aminophenoxy)-N-methyl-2-pyridinecarboxamide, 4-(4-Aminophenoxy)-N-methylpicolinamide, 4-(4-Aminophenoxy)-N-methylpyridine-2-carboxamide Imp. A (EP), 4-(4-Aminophenoxy)pyridine-2-carboxylic acid methylamide, 4-[2-(N-Methylcarbamyl)-4-pyridyloxy]aniline, 4-[[2-(Methylcarbamoyl)pyridin-4-yl]oxy]aniline, 4-[[2-(N-Methylcarbamoyl)-4-pyridyl]oxy]aniline, 4-[[2-(N-Methylcarbamoyl)pyridin-4-yl]oxy]aniline, N-Methyl-4-(4-aminophenoxy)pyridine-2-carboxamide
IUPAC Name 4-(4-aminophenoxy)-N-methylpyridine-2-carboxamide
SMILES CNC(=O)c1cc(Oc2ccc(N)cc2)ccn1
Specifications
Analyte or Component Names (4-(4-Aminophenoxy)(2-pyridyl))-N-methylcarboxamide
Shipping Condition Room Temperature
InChI Formula InChI=1S/C13H13N3O2/c1-15-13(17)12-8-11(6-7-16-12)18-10-4-2-9(14)3-5-10/h2-8H,14H2,1H3,(H,15,17)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 5 g
Formula Weight 243.1008
Chemical Name or Material (4-(4-Aminophenoxy)(2-pyridyl))-N-methylcarboxamide