missing translation for 'onlineSavingsMsg'
Learn More

4-(3,4-Dichlorophenyl)-3,4-dihydronaphthalen-1(2H)-one (Sertraline Tetralone Derivative), Mikromol™

Product Code. 30051753
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30051753

Brand: Mikromol MM0317.09

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-(3,4-Dichlorophenyl)-3,4-dihydronaphthalen-1(2H)-one (Sertraline Tetralone Derivative)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-(3',4'-Dichlorophenyl)-3,4-dihydro-2H-naphthalen-1-one
CAS 79560-19-3
Analyte or Component Names rac-(3',4'-Dichlorophenyl)-3,4-dihydro-2H-naphthalen-1-one
Shipping Condition Room Temperature
Molecular Formula C16 H12 Cl2 O
Certifications/Compliance ISO 17025
Synonym Sertraline Hydrochloride Imp. F (EP), Sertraline Imp. F (EP), 4-(3,4-Dichlorophenyl)-3,4-dihydronaphthalen-1(2H)-one, Sertraline Hydrochloride Imp. F (EP) as Racemate, Sertraline Tetralone Derivative, Sertraline Hydrochloride Impurity F as Racemate, Sertraline Impurity F as Racemate
InChI Formula InChI=1S/C16H12Cl2O/c17-14-7-5-10(9-15(14)18)11-6-8-16(19)13-4-2-1-3-12(11)13/h1-5,7,9,11H,6,8H2
SMILES Clc1ccc(cc1Cl)C2CCC(=O)c3ccccc23
IUPAC Name 4-(3,4-dichlorophenyl)-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol) 291.17
Quantity 100 mg
Formula Weight 290.0265 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.