missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Tetrazol-5-yl)benzyl alcohol, 97%, Thermo Scientific™

Product Code. 15453577
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15453577 250 mg 250mg
11411580 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15453577 Supplier Thermo Scientific Alfa Aesar Supplier No. H26964.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 501126-02-9
Molecular Formula C8H8N4O
Molecular Weight (g/mol) 176.179
MDL Number MFCD09909570
InChI Key YRBYIQONKVQONA-UHFFFAOYSA-N
Synonym 4-1h-tetrazol-5-yl benzyl alcohol, 4-1h-tetrazol-5-yl phenyl methanol, 4-1h-1,2,3,4-tetrazol-5-yl phenyl methanol, 4-1h-tetrazole-5-yl benzenemethanol, 4-2h-tetrazol-5-yl phenyl methanol, 5-4-hydroxymethylphenyl-1h-tetrazole, 4-2h-1,2,3,4-tetrazol-5-yl phenyl methanol
PubChem CID 22633334
IUPAC Name [4-(2H-tetrazol-5-yl)phenyl]methanol
SMILES C1=CC(=CC=C1CO)C2=NNN=N2

Specifications

Quantity 250 mg
Formula Weight 176.18
Percent Purity 97%
Chemical Name or Material 4-(1H-Tetrazol-5-yl)benzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.