accessibility menu, dialog, popup

UserName

OHNNHNN

CAS RN 501126-02-9

IUPAC Name: [4-(2H-1,2,3,4-tetrazol-5-yl)phenyl]methanol
Synonyms: [4-(2H-1,2,3,4-tetrazol-5-yl)phenyl]methanol
Molecular Weight (g/mol): 176.18
Molecular Formula: C8H8N4O
InChi Key: YRBYIQONKVQONA-UHFFFAOYSA-N
SMILES: OCC1=CC=C(C=C1)C1=NNN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results