Learn More
(3S,7R,7aR)-5-Benzyl-2,2-dimethyl-2,3,7,7a-tetrahydroimidazo[5,1-b]thiazole-3,7-dicarboxylic Acid, TRC
Product Code. 30424054
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30424054 25 mg
30420588 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30424054 Supplier Toronto Research Chemicals Supplier No. TRCT29346525MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2929254-14-6
Molecular Formula C16H18N2O4S
Molecular Weight (g/mol) 334.39
IUPAC Name (3S,7R,7aR)-5-benzyl-2,2-dimethyl-2,3,7,7a-tetrahydroimidazo[5,1-b]thiazole-3,7-dicarboxylic acid
SMILES O=C([C@@H]1N(C(CC2=CC=CC=C2)=N[C@@H]3C(O)=O)[C@]3([H])SC1(C)C)O
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C16H18N2O4S/c1-16(2)12(15(21)22)18-10(8-9-6-4-3-5-7-9)17-11(14(19)20)13(18)23-16/h3-7,11-13H,8H2,1-2H3,(H,19,20)(H,21,22)/t11-,12-,13+/m0/s1
Formula Weight 334.1
Chemical Name or Material (3S,7R,7aR)-5-Benzyl-2,2-dimethyl-2,3,7,7a-tetrahydroimidazo[5,1-b]thiazole-3,7-dicarboxylic Acid