CAS RN 2929254-14-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(3S,7R,7aR)-5-Benzyl-2,2-dimethyl-2,3,7,7a-tetrahydroimidazo[5,1-b]thiazole-3,7-dicarboxylic Acid, TRC
CAS: 2929254-14-6 Molecular Formula: C16H18N2O4S Molecular Weight (g/mol): 334.39 IUPAC Name: (3S,7R,7aR)-5-benzyl-2,2-dimethyl-2,3,7,7a-tetrahydroimidazo[5,1-b]thiazole-3,7-dicarboxylic acid SMILES: O=C([C@@H]1N(C(CC2=CC=CC=C2)=N[C@@H]3C(O)=O)[C@]3([H])SC1(C)C)O