Learn More
(3R,4R,6R)-5,5-Difluorohexahydro-4-hydroxy-3,6-Epoxy-2H,8H-pyrimido[6,1-b][1,3]oxazocine-8,10(9H)-di, Mikromol™
Description
(3R,4R,6R)-5,5-Difluorohexahydro-4-hydroxy-3,6-Epoxy-2H,8H-pyrimido[6,1-b][1,3]oxazocine-8,10(9H)-di
Specifications
Specifications
| Chemical Name or Material | (3R,4R,6R)-5,5-Difluorohexahydro-4-hydroxy-3,6-Epoxy-2H,8H-pyrimido[6,1-b][1,3]oxazocine-8,10(9H)-dione |
| CAS | 294177-31-4 |
| Analyte or Component Names | (3R,4R,6R)-5,5-Difluorohexahydro-4-hydroxy-3,6-Epoxy-2H,8H-pyrimido[6,1-b][1,3]oxazocine-8,10(9H)-dione |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H10 F2 N2 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | (3R,4R,6R)-5,5-Difluorohexahydro-4-hydroxy-3,6-Epoxy-2H,8H-pyrimido[6,1-b][1,3]oxazocine-8,10(9H)-dione (9CI), (4R,6R)-5,5-difluoro-4-hydroxyhexahydro-2H,8H-3,6-epoxypyrimido[6,1-b][1,3]oxazocine-8,10(9H)-dione |
| InChI Formula | InChI=1S/C9H10F2N2O5/c10-9(11)6(15)3-2-17-5-1-4(14)12-8(16)13(5)7(9)18-3/h3,5-7,15H,1-2H2,(H,12,14,16)/t3-,5?,6-,7-/m1/s1 |
| SMILES | O[C@@H]1[C@H]2COC3CC(=O)NC(=O)N3[C@H](O2)C1(F)F |
| Molecular Weight (g/mol) | 264.18 |
| Show More |