missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3R)-1-Azabicyclo[2.2.2]octan-3-ol ((3R)-3-Quinuclidinol), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
(3R)-1-Azabicyclo[2.2.2]octan-3-ol ((3R)-3-Quinuclidinol)
Specifications
Specifications
| Chemical Name or Material | (R)-(-)-3-Quinuclidinol |
| CAS | 25333-42-0 |
| Analyte or Component Names | (R)-(-)-3-Quinuclidinol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H13 N O |
| Certifications/Compliance | ISO 17025 |
| Synonym | (3R)-1-Azabicyclo[2.2.2]octan-3-ol (3-Quinuclidinol), 1-Azabicyclo[2.2.2]octan-3-ol, (R)-, 3-Quinuclidinol, (-)- (8CI), (-)-(R)-3-Quinuclidinol, (-)-3-Quinuclidinol, (3R)-1-Azabicyclo[2.2.2]octan-3-ol, (3R)-3-Quinuclidinol, (3R)-Quinuclidin-3-ol, (3R)-Quinuclidinol, (R)-(-)-1-Azabicyclo[2.2.2]octan-3-ol, (R)-(-)-3-Quinuclidinol, (R)-1-Azabicyclo[2.2.2]octan-3-ol, (R)-3-Hydroxyquinuclidine, (R)-3-Quinuclidinol, (R)-3-Quinuclidol, R-Quinuclidin-3-ol, l-3-Quinuclidinol, Solifenacin Succinate Imp. E (EP), Solifenacin Succinate Impurity E, Solifenacin Impurity E |
| InChI Formula | InChI=1S/C7H13NO/c9-7-5-8-3-1-6(7)2-4-8/h6-7,9H,1-5H2/t7-/m0/s1 |
| SMILES | O[C@H]1CN2CCC1CC2 |
| IUPAC Name | (3R)-1-azabicyclo[2.2.2]octan-3-ol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction